Learn More
Macherey-Nagel SCREW CLOSURE BONDED N 9 P
Catalog No. NC3450117
Encompass_Preferred
Screw closure (bonded), N 9, PP,blue,center hole,Silic. beige/PTFE white, 1.3 mm
Catalog No. NC3450117 Supplier Macherey-Nagel Supplier No. 702026
Screw closure (bonded), N 9, PP,blue,center hole,Silic. beige/PTFE white, 1.3 mm