Learn More
Braintree Scientific Screw Fill Vaporizer for Sevoflurane
SDP
Catalog No. 501956099
Encompass
Screw Fill Vaporizer is calibrated .25 to 5% using a laser refractometer for the specified anesthetic drug. Complete with safety lock to help prevent accidental turn-on of dial. Fluctuations in temperature, flow rate and duration of use is automatically compensated for during use. When used with I.P.P.V. the variation in output is negligible. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6099 Supplier Braintree Scientific Supplier No. VPS
Screw Fill Vaporizer is calibrated .25 to 5% using a laser refractometer for the specified anesthetic drug. Complete with safety lock to help prevent accidental turn-on of dial. Fluctuations in temperature, flow rate and duration of use is automatically compensated for during use. When used with I.P.P.V. the variation in output is negligible. **Sold for research purposes only and not for human use**