Learn More
Glass Expansion Screw Kit for Sampler Cone (PKT 4) Shimadzu 2030/2040/2050 ICP-MS (022-27001-00)
SDP
Catalog No. 502702939
Encompass_Preferred
Screw Kit for Sampler Cone (PKT 4). Package of 4 replacement screws for Shimadzu 2030 ICP-MS sampler cone. Replace sampler cone screws if corroded or worn. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2939 Supplier Glass Expansion Supplier No. SZ5004
Screw Kit for Sampler Cone (PKT 4). Package of 4 replacement screws for Shimadzu 2030 ICP-MS sampler cone. Replace sampler cone screws if corroded or worn. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001