Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Screw Mount Cable Tie

Catalog No. 19071034 Shop All 3M Products
Change view
Click to view available options
Quantity:
100/Bag
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-071-034 100/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-071-034 Supplier 3M™ Supplier No. 7000133453
  • $132.00
    $91.65 / Case of 1000

    Save $40.35

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Screw Mount Cable Tie allows controlled positioning of cables.

  • Secure wire bundles and harness components quickly, without slipping
  • Can be quickly mounted to walls and structures with screws, bolts or rivets for added security
  • Easy-to-use ties
  • High-quality 6/6 nylon
  • UL Listed 62275 Type 21 and Plenum Rated AH-2 for use in air handling spaces
  • UL Listed, RoHS 2011/65/EU Compliant and Plenum-rated
  • Comes 100/bag and 1000/case
TRUSTED_SUSTAINABILITY

Specifications

Product Type Standard Cable Tie
Certifications/Compliance UL Listed 62275 Type 21
Model CT6BK40-C
Color Black
Quantity 100/Bag
For Use With (Application) Electrical Construction and Maintenance, Mining and Metallurgy
Material Nylon 6/6
Tensile Strength (English) 40 lb.
Length (Metric) 147.32 mm
Width (Metric) 3.6 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.