Learn More
Macherey-Nagel SCREW NECK VIAL, N 9, 11.6X32.
Catalog No. NC2080978
Encompass_Preferred
Screw neck vial, N 9, 11.6x32.0 mm, 1.5 mL, label, flat bottom, amber, silanized 100/cs
Catalog No. NC2080978 Supplier Macherey-Nagel Supplier No. 702079
Screw neck vial, N 9, 11.6x32.0 mm, 1.5 mL, label, flat bottom, amber, silanized 100/cs