Learn More
Metrohm USA SCREW NIPPLE
SDP
Supplier:  Metrohm USA 62730010
Encompass
Made of PP. For 6.1448.010 Septum or for mounting 6.1403.040 Drying tube or 6.0338.100 Electrode in 6.1414.030 and 6.1414.040 KF Titration vessel lids. Used for KF titrations.
Catalog No. NC0136782