Learn More
Fritsch Milling And Sizing Inc Screw-On Lid 150-500Ml Hardmetal Tungsten Carbide P-5 Pl
SDP
Catalog No. 501534588
Encompass
With the screw-on grinding bowl lid made of hardmetal tungsten carbide, you can seal the hardmetal tungsten carbide grinding bowl with 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between filling in the glove box and the mill.
Catalog No. 50-153-4588 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.6585.00
With the screw-on grinding bowl lid made of hardmetal tungsten carbide, you can seal the hardmetal tungsten carbide grinding bowl with 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between filling in the glove box and the mill.