Learn More
Fritsch Milling And Sizing Inc Screw-On Lid 150-500Ml Hardened Stainl. Steel, P-5 Pl
SDP
Catalog No. 501534585
Encompass
With the screw-on grinding bowl lid made of hardened, stainless steel, you can seal the hardened, stainless steel grinding bowl with 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between filling in the glove box and the mill.
Catalog No. 50-153-4585 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.6555.00
With the screw-on grinding bowl lid made of hardened, stainless steel, you can seal the hardened, stainless steel grinding bowl with 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between filling in the glove box and the mill.