Learn More
Beckman Coulter Screw Ring for Analytical Cell Housing (Beckman Coulter Replacement Part)
Catalog No. NC2090748
Encompass
Beckman Coulter MPN 301922 is a replacement screw ring used in analytical ultracentrifuge cell assemblies to secure the cell housing and optical windows. It is listed in Beckman Coulter analytical cell parts and replacement parts tables and appears in assembly diagrams for external-fill and flow-through cells.
  • Compatible with analytical ultracentrifuge cell assemblies, including external-fill and flow-through cells.
  • Secures the cell housing and optical window during operation.
  • Used as a replacement spare for routine maintenance and repairs.
  • Included in replacement parts lists and exploded assembly diagrams for analytical cells.
This item is not returnable. View return policy
Catalog No. NC2090748 Supplier Beckman Coulter Supplier No. 301922
Beckman Coulter MPN 301922 is a replacement screw ring used in analytical ultracentrifuge cell assemblies to secure the cell housing and optical windows. It is listed in Beckman Coulter analytical cell parts and replacement parts tables and appears in assembly diagrams for external-fill and flow-through cells.
  • Compatible with analytical ultracentrifuge cell assemblies, including external-fill and flow-through cells.
  • Secures the cell housing and optical window during operation.
  • Used as a replacement spare for routine maintenance and repairs.
  • Included in replacement parts lists and exploded assembly diagrams for analytical cells.