Learn More
Beckman Coulter Screw-ring washer, Bakelite - for analytical rotor cell assemblies (Beckman Coulter 362328)
Catalog No. NC2090749
Encompass
Replacement Bakelite screw-ring washer used to secure the screw ring and window holder in analytical rotor cell assemblies. Supplied as a small pack intended for routine maintenance, these washers help preserve window seating and rotor cell integrity.
  • Bakelite material for durable seating and stability.
  • Pack contains multiple washers for repeated replacements.
  • Secures screw ring and window holder within the cell housing.
  • Supports reliable window seating to help prevent leaks.
  • Designed for routine maintenance and repair of analytical rotor cells.
This item is not returnable. View return policy
Catalog No. NC2090749 Supplier Beckman Coulter Supplier No. 362328
Replacement Bakelite screw-ring washer used to secure the screw ring and window holder in analytical rotor cell assemblies. Supplied as a small pack intended for routine maintenance, these washers help preserve window seating and rotor cell integrity.
  • Bakelite material for durable seating and stability.
  • Pack contains multiple washers for repeated replacements.
  • Secures screw ring and window holder within the cell housing.
  • Supports reliable window seating to help prevent leaks.
  • Designed for routine maintenance and repair of analytical rotor cells.