Learn More
JG FINNERAN SCREW THREAD CLOSURE 1000/CS
Catalog No. NC2231303
Encompass_Preferred
JG Finneran 806550-13 Preassembled Screw Thread Closure with PTFE/Silicone Septum, 0.065" Thick, Black (Case of 1000)
This item is not returnable. View return policy
Catalog No. NC2231303 Supplier JG FINNERAN Supplier No. 80655013
JG Finneran 806550-13 Preassembled Screw Thread Closure with PTFE/Silicone Septum, 0.065" Thick, Black (Case of 1000)