Learn More
StatLab Screw Top Empty Specimen Containers
Description
- StatLab containers are designed for safe, leak-resistant transport and storage of fluids
- Screw-top containers have knurled cap and base for easy handling
- They are graduated and made of durable clarified polypropylene
- Lids included
Specifications
Specifications
| Product Type | Specimen Container |
| Closure Included | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Capacity (Metric) | 500 mL |