Learn More
Uline Screw Top Pail - 6.5 Gallon, White Lid
Catalog No. NC3836055
Encompass
Quickly open and reseal for easy access.Vacuum tight – Use with granulars, fine powders.Lid includes neoprene gasket for moisture-resistant storage.Tamper evident with use of cable ties.Molded, high-density polyethylene.FDA compliant.
Catalog No. NC3836055 Supplier Uline Supplier No. S15637W
Quickly open and reseal for easy access.Vacuum tight – Use with granulars, fine powders.Lid includes neoprene gasket for moisture-resistant storage.Tamper evident with use of cable ties.Molded, high-density polyethylene.FDA compliant.