Learn More
Thermo Scientific™ Screw Vial Convenience Kit, solid-top cap, 4ml clear glass vial
Catalog No. 033917B Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
03-391-7B Clear
1 options
This item is not returnable. View return policy
Catalog No. 03-391-7B Supplier Thermo Scientific™ Supplier No. B78002
This item is not returnable. View return policy
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
TRUSTED_SUSTAINABILITY
Specifications
Description Screw Vial Convenience Kit, Solid-top Cap
Color Clear
Material Glass
Volume (Metric) 4 mL
No. per Pack 100
Closure Type Screw Cap, Solid Top