Learn More
Microsolv Technology Corporation Screwcap 4Ml Sil/Ptfe Bonded
Catalog No. 5001452231
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Septa, bonded in knurled, polypropylene, Black Caps. For use with 13-425mm thread, 15x45mm autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52231 Supplier Microsolv Technology Corporation Supplier No. 9504S10CBK
Caps, Screw Top, with Silicone Rubber / PTFE Septa, bonded in knurled, polypropylene, Black Caps. For use with 13-425mm thread, 15x45mm autosampler vials. MicroSolv Brand. 100/PK.