Learn More
Microsolv Technology Corporation Screwcap 4Ml Slit Sil/Ptf Bndd
Catalog No. 5001452233
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Black Caps. For use with 13-425mm thread, 15x45mm autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52233 Supplier Microsolv Technology Corporation Supplier No. 9504S30CBK
Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Black Caps. For use with 13-425mm thread, 15x45mm autosampler vials. MicroSolv Brand. 100/PK.