Learn More
Beckman Coulter Beckman Coulter Screwdriver for Housing Plug - Flat-edge tool for rotor cell filling holes
Catalog No. NC2882985
Encompass
The Beckman Coulter Screwdriver for Housing Plug (MPN 807371) is a specialized flat-edge tool used to install and remove housing plugs in rotor cell filling holes on analytical and preparative ultracentrifuge rotors. It aids precise seating and removal of plugs to ensure rotor cell integrity and safe operation.
  • Specialized flat-edge tip for engaging housing plugs.
  • Designed for seating and removing housing plugs in filling holes.
  • Compact handheld tool for precise control during assembly and maintenance.
  • Durable construction to withstand repeated use in lab environments.
  • Suitable for use with analytical and preparative rotor cells and assemblies.
Catalog No. NC2882985 Supplier Beckman Coulter Supplier No. 807371
The Beckman Coulter Screwdriver for Housing Plug (MPN 807371) is a specialized flat-edge tool used to install and remove housing plugs in rotor cell filling holes on analytical and preparative ultracentrifuge rotors. It aids precise seating and removal of plugs to ensure rotor cell integrity and safe operation.
  • Specialized flat-edge tip for engaging housing plugs.
  • Designed for seating and removing housing plugs in filling holes.
  • Compact handheld tool for precise control during assembly and maintenance.
  • Durable construction to withstand repeated use in lab environments.
  • Suitable for use with analytical and preparative rotor cells and assemblies.