Learn More
Medchemexpress LLC HY-15489 10mg Medchemexpress, Scriptaid CAS:287383-59-9 Purity:>98%
SDP
Catalog No. 502030637
Encompass_Preferred
Medchemexpress, HY-15489 10mg Scriptaid CAS:287383-59-9 Scriptaid is a potent histone deacetylase (HDAC) inhibitor, used in cancer research. Scriptaid is also a sensitizer to antivirals and has potential for epstein-barr virus (EBV)-associated lymphomas treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0637 Supplier Medchemexpress LLC Supplier No. HY1548910MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-15489 10mg Scriptaid CAS:287383-59-9 Scriptaid is a potent histone deacetylase (HDAC) inhibitor, used in cancer research. Scriptaid is also a sensitizer to antivirals and has potential for epstein-barr virus (EBV)-associated lymphomas treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.