Learn More
Enzo Life Sciences Scriptaid (1mg). CAS: 287383-59-9
SDP
Supplier:  Enzo Life Sciences BMLGR3260001
Encompass_Preferred
Inhibitor of histone deacetylase (HDAC) (HDAC1 and 3: IC50~0.6µM; HDAC8: IC50~1µM) with lower toxicity than trichostatin A (BML-GR309). Inhibits tumor growth in vitro and in vivo and, in conjunction with 5-aza-2’-deoxycytidine (ALX-480-096), enhances expression of estrogen receptor α. Optimal concentration 2-2.5μg/ml (6-8μM). A negative control is available (BML-GR327). Alternative name: 6-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxyhexanamide. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (4mg/ml), ethanol (0.3mg/ml) or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0980