Learn More
Cellscript Inc ScriptCap™ Cap 1 Capping System, 50 reactions, post-transcriptional 5' capping, Cap 0 or Cap 1, nearly 100% capping efficiency, 45 min reaction time, 60 μg RNA capacity
Catalog No. NC2953874
Encompass
ScriptCap™ Cap 1 Capping System, 50 reactions, post-transcriptional 5' capping, Cap 0 or Cap 1, nearly 100% capping efficiency, 45 min reaction time, 60 μg RNA capacity
Catalog No. NC2953874 Supplier Cellscript Inc Supplier No. CSCCS2250
ScriptCap™ Cap 1 Capping System, 50 reactions, post-transcriptional 5' capping, Cap 0 or Cap 1, nearly 100% capping efficiency, 45 min reaction time, 60 μg RNA capacity