Learn More
Cellscript Inc ScriptGuard RNase Inhibitor, 40 U/μl, 10,000 units, 1:1 binding ratio, Kᵢ ~10⁻¹⁴ M, storage -20 °C, 50% glycerol solution
Supplier:  Cellscript Inc CSRI6310K
Encompass
ScriptGuard RNase Inhibitor, 40 U/μl, 10,000 units, 1:1 binding ratio, Kᵢ ~10⁻¹⁴ M, storage -20 °C, 50% glycerol solution
Catalog No. NC2039205