Learn More
Med Vet International E-Z Scrub Brush, 1% Povidone Iodine, 30/Packg
Catalog No. 502831129
Encompass_Preferred
Single use disposable surgical scrub brushes impregnated with a wide variety of antimicrobial agents, including chlorhexidine gluconate (CHG), parachlorometaxylenol (PCMX) and povidone iodine (PVP-I). Proprietary polyethylene soft bristles enhance mechanical action while reducing skin irritation. Soft sponge provides gentle cleaning on irritated areas or when firm mechanical action is not required. Ergonomic brush design allows greater user comfort and effectiveness, ultimately improving compliance. Single packaged brushes.
Catalog No. 50-283-1129 Supplier Med Vet International Supplier No. 372053
Single use disposable surgical scrub brushes impregnated with a wide variety of antimicrobial agents, including chlorhexidine gluconate (CHG), parachlorometaxylenol (PCMX) and povidone iodine (PVP-I). Proprietary polyethylene soft bristles enhance mechanical action while reducing skin irritation. Soft sponge provides gentle cleaning on irritated areas or when firm mechanical action is not required. Ergonomic brush design allows greater user comfort and effectiveness, ultimately improving compliance. Single packaged brushes.