Learn More
Chemglass Life Sciences Scrubber/Absorber Tube Only, Arsine Generator
SDP
Catalog No. CG109002
Encompass_Preferred
Scrubber/Absorber Tube Only, Arsine Generator. Replacement scrubber/absorber tube for item CG109001. Complete item CG109001 consists of a 125mL Erlenmeyer flask and scrubber tube both having 24/40 standard taper joints. Also supplied complete with one CG-145-05, size 24 Keck clip. Glass beads may be added to the absorber tube to enhance the gas mixing.
Catalog No. CG109002 Supplier Chemglass Life Sciences Supplier No. CG109002
Scrubber/Absorber Tube Only, Arsine Generator. Replacement scrubber/absorber tube for item CG109001. Complete item CG109001 consists of a 125mL Erlenmeyer flask and scrubber tube both having 24/40 standard taper joints. Also supplied complete with one CG-145-05, size 24 Keck clip. Glass beads may be added to the absorber tube to enhance the gas mixing.