Learn More
MSC AbilityOne Floor Pad, Deep Scrubbing, Round, 17 in, Blue
Catalog No. 502839522
Encompass
This floor pad is designed for heavy-duty scrubbing applications, effectively removing dirt and scuff marks from floors prior to recoating. Made from non-woven polyester, it features a round shape and a blue color, suitable for use with rotary or automatic floor machines.
  • Designed for deep scrubbing
  • Round shape
  • 17 inch diameter
  • Constructed from non-woven polyester material
  • Blue color for easy identification
  • Compatible with rotary or automatic floor machines
  • Effectively removes dirt and scuff marks
Catalog No. 50-283-9522 Supplier MSC Supplier No. 89466510
This floor pad is designed for heavy-duty scrubbing applications, effectively removing dirt and scuff marks from floors prior to recoating. Made from non-woven polyester, it features a round shape and a blue color, suitable for use with rotary or automatic floor machines.
  • Designed for deep scrubbing
  • Round shape
  • 17 inch diameter
  • Constructed from non-woven polyester material
  • Blue color for easy identification
  • Compatible with rotary or automatic floor machines
  • Effectively removes dirt and scuff marks