Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ Scrubbing Towels
SDP

Catalog No. 19043853 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Quantity:
170 Wipes/Canister
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-043-853 170 Wipes/Canister
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-853 Supplier GOJO™ Supplier No. 639802
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

No water needed towels for use anywhere heavy duty cleaning is required. Dual textured wet towel for quick and convienient cleaning.

  • Dual textured with a scrubbing side for removing tough soils and a smooth side for quick dirt absorption
  • Extra-large towels for superior cleaning capacity
  • Contains skin conditioners to help prevent dryness and irritation
  • For use anywhere heavy duty cleaning is required
  • No water needed
  • Fresh citrus scent
  • Effective on oil, grease, paint, adhesives, tar, and asphalt
TRUSTED_SUSTAINABILITY

Specifications

Product Type Heavy Duty Hand Cleanser
Color Gray
Form Wipe
For Use With (Application) Effective on oil, grease, paint, adhesives, tar, and asphalt.
For Use With (Equipment) 6302-01
Packaging Bucket
Quantity 170 Wipes/Canister
Size 170 count
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.