Learn More
MSC PortionPac Heavy-Duty Degreaser, Water Based, Automatic Scrubbers, 216/Pk.
Catalog No. 502868487
Encompass
This heavy-duty degreaser is designed for effective floor cleaning, delivered in a convenient 3 fl oz pouch. Its water-based, ultra-concentrated formula is optimized for use with automatic scrubbers.
  • Features a pH of 9.90
  • Ultra-concentrated solution
  • Green Seal certified
  • Biodegradable formulation
  • Water-based for automatic scrubbers
Catalog No. 50-286-8487 Supplier MSC Supplier No. 41752627
This heavy-duty degreaser is designed for effective floor cleaning, delivered in a convenient 3 fl oz pouch. Its water-based, ultra-concentrated formula is optimized for use with automatic scrubbers.
  • Features a pH of 9.90
  • Ultra-concentrated solution
  • Green Seal certified
  • Biodegradable formulation
  • Water-based for automatic scrubbers