Learn More
MSC Nilfisk Squeegee Carpet Kit, 321 mm
Catalog No. 502836400
Encompass
The Nilfisk 321 mm Carpet Squeegee Kit is an essential accessory designed for efficient water extraction from carpets. It is compatible with SC100 and MA10 upright scrubbers, enabling these machines to scrub and dry carpeted areas in a single pass, enhancing cleaning performance.
  • Designed for efficient water extraction from carpets.
  • Compatible with SC100 upright scrubbers.
  • Compatible with MA10 upright scrubbers.
  • Facilitates scrubbing and drying in a single pass.
  • Measures 321 mm in length.
Catalog No. 50-283-6400 Supplier MSC Supplier No. 35632454
The Nilfisk 321 mm Carpet Squeegee Kit is an essential accessory designed for efficient water extraction from carpets. It is compatible with SC100 and MA10 upright scrubbers, enabling these machines to scrub and dry carpeted areas in a single pass, enhancing cleaning performance.
  • Designed for efficient water extraction from carpets.
  • Compatible with SC100 upright scrubbers.
  • Compatible with MA10 upright scrubbers.
  • Facilitates scrubbing and drying in a single pass.
  • Measures 321 mm in length.