Learn More
MSC SCRUBS Hand Cleaner Towel, Pre-Moistened, 30 Count, 10 x 12 in, Citrus Scent
Catalog No. 502836960
Encompass
SCRUBS in-a-Bucket Hand Cleaner Towels are pre-moistened wipes that combine an advanced cleaning formula with a strong, dual-textured towel to thoroughly clean hands and surfaces. These alcohol-free, citrus-scented towels effectively remove industrial soils like dirt, tar, and grease, leaving no sticky residue and requiring no rinsing.
  • Advanced cleaning power for hands and surfaces
  • Leaves no sticky residue
  • Thoroughly cleans hands and surfaces anywhere, anyplace, anytime!
  • Formulated to prevent hand dryness and irritation
  • Locks in soil to prevent transfer
  • No rinsing required
  • Strong, dual-textured towel construction
  • Clean, citrus fragrance
  • Alcohol-free formula
Catalog No. 50-283-6960 Supplier MSC Supplier No. 66553298
SCRUBS in-a-Bucket Hand Cleaner Towels are pre-moistened wipes that combine an advanced cleaning formula with a strong, dual-textured towel to thoroughly clean hands and surfaces. These alcohol-free, citrus-scented towels effectively remove industrial soils like dirt, tar, and grease, leaving no sticky residue and requiring no rinsing.
  • Advanced cleaning power for hands and surfaces
  • Leaves no sticky residue
  • Thoroughly cleans hands and surfaces anywhere, anyplace, anytime!
  • Formulated to prevent hand dryness and irritation
  • Locks in soil to prevent transfer
  • No rinsing required
  • Strong, dual-textured towel construction
  • Clean, citrus fragrance
  • Alcohol-free formula