Learn More
CHEMSCIENCE Screw Cap and BONDED/PRE-SLIT Septa, Size 9-425, Red PTFE/White Silicone 1mm(0.040),CS/1000
Catalog No. NC2681284
Encompass_Preferred
Screw Cap and BONDED/PRE-SLIT Septa, Size 9-425, Red PTFE/White Silicone 1mm(0.040), PK/100,CS/10PKS
Catalog No. NC2681284 Supplier CHEMSCIENCE Supplier No. CSCS09SBS
Screw Cap and BONDED/PRE-SLIT Septa, Size 9-425, Red PTFE/White Silicone 1mm(0.040), PK/100,CS/10PKS