Learn More
Neta Scientific Agilent Screw cap vials, pre-assembled, 100/PK
SDP
Catalog No. 50929225
Encompass_Preferred
This item is manufactured by Agilent
Agilent Vial pack, screw top, pre-assembled, clear vials with write-on spot, blue caps, certified, PTFE/red rubber septa, 2 mL, 100/pk. Vial size: 12 x 32 mm (12 mm cap) - 2 ml Screw Top Vials & Caps
UNSPSC Code  41121806
- 58
Catalog No. 50-929-225 Supplier Neta Scientific Supplier No. 51820864
This item is manufactured by Agilent
Agilent Vial pack, screw top, pre-assembled, clear vials with write-on spot, blue caps, certified, PTFE/red rubber septa, 2 mL, 100/pk. Vial size: 12 x 32 mm (12 mm cap) - 2 ml Screw Top Vials & Caps
UNSPSC Code  41121806
- 58