Learn More
Grainger Socket Set Screw, Cup, M2.5 x 0.45mm x 3mm, Pk50
Catalog No. NC2180910
Encompass_Preferred
The SCKT SET SCRW,CUP,M2.5X0.45MMX3MM,PK50 is a pack of 50 M2.5 x 0.45mm socket head screws with 3mm cup point. The screws are made of stainless steel with a tensile strength of 900 MPa. They feature a hex socket drive for use with hex keys. The screws have a thread diameter of 2.5mm and a head diameter of 4.5mm. They are 3mm in length. The screws are RoHS compliant. This pack is manufactured by SCKT and can be used for electronic applications.
This item is not returnable. View return policy
Catalog No. NC2180910 Supplier Grainger Supplier No. 6EY82
The SCKT SET SCRW,CUP,M2.5X0.45MMX3MM,PK50 is a pack of 50 M2.5 x 0.45mm socket head screws with 3mm cup point. The screws are made of stainless steel with a tensile strength of 900 MPa. They feature a hex socket drive for use with hex keys. The screws have a thread diameter of 2.5mm and a head diameter of 4.5mm. They are 3mm in length. The screws are RoHS compliant. This pack is manufactured by SCKT and can be used for electronic applications.