Learn More
Neta Scientific Agilent Vial,screw,300ul,w/insrt,clr,100PK
SDP
Catalog No. 50929241
Encompass_Preferred
This item is manufactured by Agilent
Agilent Vial, screw top, with fixed insert, clear, 300 uL insert volume, 100/pk. Vial size: 12 x 32 mm (12 mm cap) - High Recovery Vials & Inserts
UNSPSC Code  41121806
- 58
Catalog No. 50-929-241 Supplier Neta Scientific Supplier No. 51886591
This item is manufactured by Agilent
Agilent Vial, screw top, with fixed insert, clear, 300 uL insert volume, 100/pk. Vial size: 12 x 32 mm (12 mm cap) - High Recovery Vials & Inserts
UNSPSC Code  41121806
- 58