Learn More
Foxx Life Sciences EZBio Silicone Cap System, 83B Overmolded Caps, PP, with 2x1/4" Tubing for 4L HD Bottle Cap Assembly and 2.5L Foxx Rectangular Carboy Cap Assembly, Sterile, 1/EA
GREENER_CHOICE
Catalog No. 5000044045
Encompass_Preferred
EZBio Silicone Cap System is a bottle cap assembly made from overmolded platinum-cured silicone, compliant with USP Class VI standards. Designed as a cleanroom-compatible closure for pharmaceutical container closures in ISO-certified cleanroom, this joint-free system eliminates fluid entrapment & leak risk while supporting closed-loop fluid transfer, media & buffer handling, & single-use systems. Compatible with gamma sterilization & offered as a gamma-irradiated silicone closure, it’s ideal for aseptic workflows & custom bioprocessing assembly.
Available in 38-430, GL45, 53B, &83B sizes, they fit glass, plastic & stainless-steel containers.
Features
Validation package for efficient qualification
Joint-free, platinum-cured silicone caps for leak-free, contamination-resistant use
USP Class VI, non-cytotoxic materials
Translucent design for fluid path visibility
Wide temperature range: -76°F to +428°F (-60°C to +220°C)
Reduces cleaning & assembly costs
Catalog No. 50-000-44045 Supplier Foxx Life Sciences Supplier No. 2059105FLS
EZBio Silicone Cap System is a bottle cap assembly made from overmolded platinum-cured silicone, compliant with USP Class VI standards. Designed as a cleanroom-compatible closure for pharmaceutical container closures in ISO-certified cleanroom, this joint-free system eliminates fluid entrapment & leak risk while supporting closed-loop fluid transfer, media & buffer handling, & single-use systems. Compatible with gamma sterilization & offered as a gamma-irradiated silicone closure, it’s ideal for aseptic workflows & custom bioprocessing assembly.
Available in 38-430, GL45, 53B, &83B sizes, they fit glass, plastic & stainless-steel containers.
Features
Validation package for efficient qualification
Joint-free, platinum-cured silicone caps for leak-free, contamination-resistant use
USP Class VI, non-cytotoxic materials
Translucent design for fluid path visibility
Wide temperature range: -76°F to +428°F (-60°C to +220°C)
Reduces cleaning & assembly costs