Learn More
MSC SCS Smock Jacket, Knitted Cuffs, Antistatic, Groundable, 1 ea
Catalog No. 5000686720
Encompass
SCS smocks are designed to provide antistatic, low tribocharging protection, creating a Faraday Cage around the wearer. These garments protect ESD susceptible items from electrostatic fields and meet ANSI/ESD S20.20 requirements for groundable static control.
  • Panel-to-panel conductivity
  • Patented hip-to-cuff grounding
  • Jacket length with set-in style sleeves and reinforced hem
  • V-neck and stainless steel snaps
  • Tag-free neck label
  • Two inch knitted elastic conductive cuffs
  • Three pockets
  • Made in the United States of America
Catalog No. 50-006-86720 Supplier MSC Supplier No. 10913341
SCS smocks are designed to provide antistatic, low tribocharging protection, creating a Faraday Cage around the wearer. These garments protect ESD susceptible items from electrostatic fields and meet ANSI/ESD S20.20 requirements for groundable static control.
  • Panel-to-panel conductivity
  • Patented hip-to-cuff grounding
  • Jacket length with set-in style sleeves and reinforced hem
  • V-neck and stainless steel snaps
  • Tag-free neck label
  • Two inch knitted elastic conductive cuffs
  • Three pockets
  • Made in the United States of America