Learn More
Agilent Technologies SCS110 COMPETENT CELLS
Catalog No. 50125056
Encompass
SCS110 Competent Cells 5 x 0.2mL= 5 x 106 transformants/microgram Deficient in Dam and Dcm methylases DNA propagated in these strains can be digested by methylation-sensitive enzymes SCS110 strain an EndA- version of JM110
Catalog No. 50-125-056 Supplier Agilent Technologies Supplier No. 200247
SCS110 Competent Cells 5 x 0.2mL= 5 x 106 transformants/microgram Deficient in Dam and Dcm methylases DNA propagated in these strains can be digested by methylation-sensitive enzymes SCS110 strain an EndA- version of JM110