Learn More
Agilent Technologies SCSI SUPERCOMP CELLS
Catalog No. 50125048
Encompass
SCS1 cells are endonuclease (endA) deficient, Improves the quality of miniprep DNA, and are recombination (recA)deficient, improving insert stability. The hsdR mutation prevents the cleavage of cloned DNA by the EcoK endonuclease system.
Catalog No. 50-125-048 Supplier Agilent Technologies Supplier No. 200231
SCS1 cells are endonuclease (endA) deficient, Improves the quality of miniprep DNA, and are recombination (recA)deficient, improving insert stability. The hsdR mutation prevents the cleavage of cloned DNA by the EcoK endonuclease system.