Learn More
Med Vet International Miltex Iris Scissors, SuperCut, Straight, Sharp/Sharp, German, 4.62in, 5-SC-304
Catalog No. 503045615
Encompass_Preferred
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. SuperCut blades allow for easy use. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. Straight sharp/sharp pointed blades. 4.62in total length. Made with high-quality, high-end German steel.
Catalog No. 50-304-5615 Supplier Med Vet International Supplier No. MIL5SC304
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. SuperCut blades allow for easy use. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. Straight sharp/sharp pointed blades. 4.62in total length. Made with high-quality, high-end German steel.