Learn More
Med Vet International 5 1/2in. (14cm) Littauer Stitch Scissors, German Grade, Von Klaus
Catalog No. 503046050
Encompass_Preferred
Von Klaüs German grade. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6050 Supplier Med Vet International Supplier No. VK1140614
Von Klaüs German grade. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.