Learn More
Med Vet International Miltex MeisterHand Stevens Tenotomy Scissors, Straight, Short Blades, Sharp Points, German, 4.25in, MH18-1460
Catalog No. 503045754
Encompass_Preferred
Typically used in the optical field, tenotomy scissors are mainly used in delicate surgical procedures. They are especially used in ophthalmic and neurosurgery. MeisterHand scissors guarantee signiture exceptional craftsmanship. The small and sharp blades allows for precision cutting of the sensitive and delicate tissues in the eye region and of the small nerves. The blades are straight, short blades culminating into sharp tips. 4.25in in length. These scissors are made with high-quality, medical-grade German steel.
Catalog No. 50-304-5754 Supplier Med Vet International Supplier No. MILMH181460
 Restricted Item
Add to Cart
Add to Cart
Typically used in the optical field, tenotomy scissors are mainly used in delicate surgical procedures. They are especially used in ophthalmic and neurosurgery. MeisterHand scissors guarantee signiture exceptional craftsmanship. The small and sharp blades allows for precision cutting of the sensitive and delicate tissues in the eye region and of the small nerves. The blades are straight, short blades culminating into sharp tips. 4.25in in length. These scissors are made with high-quality, medical-grade German steel.