Learn More
Med Vet International Utility Scissors, 6.5", Blunt/Blunt Tip, German Grade, Von Klaus
Catalog No. 503046040
Encompass_Preferred
6 1/2in. German Grade extra heavy utility scissors have 1 serrated blade and a blunt/blunt tip. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6040 Supplier Med Vet International Supplier No. VK1031702
6 1/2in. German Grade extra heavy utility scissors have 1 serrated blade and a blunt/blunt tip. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.