Learn More
Med Vet International Miltex Blepharoplasty Scissors, Curved, One Serrated Blade, Ribbon Type, German, 4.12in, 5-263
Catalog No. 503046737
Encompass_Preferred
Typically used to cut eye tissue for upper and lower blepharoplasty. The serrated blade ensures optimal grip of tissue, preventing slips. 4.12in in length. Curved blades with a serrated finish. Made with high-end, medical grade German steel.
Catalog No. 50-304-6737 Supplier Med Vet International Supplier No. MIL5263
Typically used to cut eye tissue for upper and lower blepharoplasty. The serrated blade ensures optimal grip of tissue, preventing slips. 4.12in in length. Curved blades with a serrated finish. Made with high-end, medical grade German steel.