Learn More
Aviva Systems Biology Corp SCTR Antibody - N-terminal region (ARP65498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCTR. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human SCTR. Alternate names: SRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human
Catalog No. 50-288-509