Learn More
Ace Glass, Inc. Septa, Suba-Seal, white, #25 fits 14mm ID necks, fits 14/20 joints, pk/100
SDP
Catalog No. NC0480077
Encompass
#25 Suba-Seal white rubber septa designed with serrated bottom for sealing open-top glass vessels with 14mm I.D. necks, as well as, 14/20 standard taper joints. Pack/100. Ideal for moisture or air-sensitive applications. Autoclavable.
Catalog No. NC0480077 Supplier Ace Glass, Inc. Supplier No. 9107123
#25 Suba-Seal white rubber septa designed with serrated bottom for sealing open-top glass vessels with 14mm I.D. necks, as well as, 14/20 standard taper joints. Pack/100. Ideal for moisture or air-sensitive applications. Autoclavable.