Learn More
Medchemexpress LLC Scutebarbatine A | 176520-13-1 | 558.62 | 1 MG
SDP
Catalog No. 5000416482
Encompass_Preferred
Scutebarbatine A is a chemical substance used as a laboratory chemical for research purposes. It inhibits the proliferation of HCC cells and induces apoptosis by activating the MAPK and ER stress signaling pathways. This product is not intended for medical applications and is for research use only.
  • Inhibits proliferation of HCC cells.
  • Triggers apoptosis via activation of MAPK and ER stress.
  • Appears as a white to off-white solid.
  • Classified as a natural product.
  • Belongs to the alkaloid and pyridine alkaloid classifications.
Catalog No. 50-004-16482 Supplier Medchemexpress LLC Supplier No. HYN12601MG
Scutebarbatine A is a chemical substance used as a laboratory chemical for research purposes. It inhibits the proliferation of HCC cells and induces apoptosis by activating the MAPK and ER stress signaling pathways. This product is not intended for medical applications and is for research use only.
  • Inhibits proliferation of HCC cells.
  • Triggers apoptosis via activation of MAPK and ER stress.
  • Appears as a white to off-white solid.
  • Classified as a natural product.
  • Belongs to the alkaloid and pyridine alkaloid classifications.