Learn More
Cayman Chemical Scutellarein, Purity: ≥98%, CAS: 529-53-3, 10 mg
SDP
Catalog No. 502935129
Encompass_Preferred
A flavone; has antioxidant activity in a TRAP assay at 1 μg/ml; inhibits sucrose hydrolysis by rat intestinal α-glucosidase and ATPase activity of the SARS coronavirus helicase nsP13 (IC50s = 12 and 0.86 μM, respectively); decreases LPS-induced NO release and iNOS and TNF-α mRNA expression in RAW 264.7 macrophages at 50 μM; decreases tumor weight and volume in an HT-1080 human fibrosarcoma mouse xenograft model at 0.5 μg/g
Catalog No. 50-293-5129 Supplier Cayman Chemical Supplier No. 2497710 MG
A flavone; has antioxidant activity in a TRAP assay at 1 μg/ml; inhibits sucrose hydrolysis by rat intestinal α-glucosidase and ATPase activity of the SARS coronavirus helicase nsP13 (IC50s = 12 and 0.86 μM, respectively); decreases LPS-induced NO release and iNOS and TNF-α mRNA expression in RAW 264.7 macrophages at 50 μM; decreases tumor weight and volume in an HT-1080 human fibrosarcoma mouse xenograft model at 0.5 μg/g