Learn More
Medchemexpress LLC HY-N4314 1mg Medchemexpress, Scutellarein tetramethyl ether CAS:1168-42-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN43141MG
Encompass_Preferred
Medchemexpress, HY-N4314 1mg Scutellarein tetramethyl ether CAS:1168-42-9 Scutellarein tetramethyl ether (4',5,6,7-Tetramethoxyflavone) is a bioactive component of Siam weed extract. Scutellarein tetramethyl ether (4',5,6,7-Tetramethoxyflavone) exhibits anti-inflammatory activity through NF-κB pathway [1] . Scutellarein tetramethyl ether (4',5,6,7-Tetramethoxyflavone) modulats of bacterial drug resistance via efflux pump inhibition [2] . Scutellarein tetramethyl ether (4',5,6,7-Tetramethoxyflavone) can enhance blood coagulation [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3952