Learn More
Medchemexpress LLC Scutellarein tetramethyl ether | 1168-42-9 | MFCD00017442 | 99.9% | 342.34 g/mol | C19H18O6 | 10 MG
SDP
Catalog No. 5000289944
Encompass_Preferred
Scutellarein tetramethyl ether is a flavone derivative (4',5,6,7-tetramethoxyflavone) provided as a research-grade solid for biochemical and cellular studies. It has reported anti-inflammatory, antibacterial, pro-coagulant, and anti-tumor activities and is used in assays probing NF-κB signaling, bacterial efflux pump function, coagulation pathways, and cancer cell growth inhibition.
  • High purity solid suitable for biochemical and cellular assays.
  • Molecular weight 342.34 g/mol and formula C19H18O6.
  • Reported anti-inflammatory, antibacterial, pro-coagulant, and anti-tumor activities.
  • Available in small pack sizes for screening and research.
  • Provided with a datasheet containing CAS, purity, and handling information.
Catalog No. 50-002-89944 Supplier Medchemexpress LLC Supplier No. HYN431410MG
Scutellarein tetramethyl ether is a flavone derivative (4',5,6,7-tetramethoxyflavone) provided as a research-grade solid for biochemical and cellular studies. It has reported anti-inflammatory, antibacterial, pro-coagulant, and anti-tumor activities and is used in assays probing NF-κB signaling, bacterial efflux pump function, coagulation pathways, and cancer cell growth inhibition.
  • High purity solid suitable for biochemical and cellular assays.
  • Molecular weight 342.34 g/mol and formula C19H18O6.
  • Reported anti-inflammatory, antibacterial, pro-coagulant, and anti-tumor activities.
  • Available in small pack sizes for screening and research.
  • Provided with a datasheet containing CAS, purity, and handling information.