Learn More
TARGETMOL CHEMICALS INC Scutellarein tetramethyl ether
SDP
Catalog No. 502880258
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Scutellarein tetramethyl ether (4',5,6,7-Tetramethoxyflavone) is extracted from Siam weeds and demonstrates anti-inflammatory activity via the NF-kB pathway. Purity 98.43%
Catalog No. 50-288-0258 Supplier TARGETMOL CHEMICALS INC Supplier No. TN12715MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Scutellarein tetramethyl ether (4',5,6,7-Tetramethoxyflavone) is extracted from Siam weeds and demonstrates anti-inflammatory activity via the NF-kB pathway. Purity 98.43%