Learn More
Selleck Chemical LLC Scutellarin 10mg 27740-01-8 Breviscapine, Breviscapin, Scutellarein-7-glucuronide
SDP
Supplier:  Selleck Chemical LLC S381010MG
Encompass_Preferred
Scutellarin (Breviscapine, Breviscapin, Scutellarein-7-glucuronide), the major active principal flavonoids extracted from the Chinese herbal medicines Scutellaria baicalensis and Erigeron breviscapus (Vant.) Hand-Mazz, has many pharmacological effects, such as antioxidant, antitumor, antiviral, and antiinflammatory activities. Scutellarin can down-regulates the STAT3/Girdin/Akt signaling in HCC cells, and inhibits RANKL-mediated MAPK and NF-?B signaling pathway in osteoclasts. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8440