Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scutellarin, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013802 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3802 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3802 Supplier MilliporeSigma Supelco Supplier No. 7357710MG
Add to Cart
Add to Cart

Scutellarin is flavone glucuronide and an active ingredient of breviscapine. It is widely used for the treatment of cerebral infarction, coronary heart disease and angina pectoris.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 27740-01-8
Physical Form Neat
Linear Formula C21H18O12
MDL Number MFCD01861503
Synonym 4 ′,5,6,7-Tetrahydroxyflavone 7-glucuronide; 5,6-Dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4 H-1-benzopyran-7-yl-beta-D-glucopyranosiduronic acid; 7-(beta-D-Glucopyranuronosyloxy)-5,6-dihydroxy-2-(4-hydroxyphenyl)-4 H-1-benzopyran-4-one; Breviscapin; Scutellarein-7-O-beta-D-glucuronide
Type Analytical Standard
Quantity 10 mg
Formula Weight 462.36
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.